Assay Details
Target Gene Details
Entrez Gene ID: | 16912 |
Gene Name: | proteasome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2) |
Gene Aliases: |
Lmp-2, Lmp2 |
Location: |
Chr.17:34182030-34187330 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmb9 | NM_013585.2 | NP_038613.1 | ||
AK008429.1 | BAB25664.1 | |||
AK152743.1 | BAE31463.1 | |||
AK171670.1 | BAE42601.1 | |||
BC116364.2 | ||||
CT010227.1 | CAJ18435.1 | |||
D44454.1 | BAA22575.1 | |||
D44456.1 | BAA22577.1 | |||
D44457.1 | BAA22578.1 | |||
D44458.1 | BAA22579.1 | |||
D44461.1 | BAA22582.1 | |||
KJ650226.1 | ||||
S58203.1 | AAP13903.1 | |||
S59862.1 | AAB20105.1 | |||
U22447.1 | AAA75304.1 | |||
U22448.1 | AAA75305.1 | |||
U22919.1 | AAA75306.1 | |||
U22920.1 | AAA75307.1 |
More Information
Set Membership: |
Intragenic Non-exonic |