Assay Details
Target Gene Details
Entrez Gene ID: | 80905 |
Gene Name: | polymerase (DNA directed), eta (RAD 30 related) |
Gene Aliases: |
RAD30A, XPV |
Location: |
Chr.17:46171355-46202683 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polh | NM_030715.3 | 8 | 1176 | NP_109640.1 |
AB027128.1 | 8 | 1202 | BAA97570.1 | |
AK036296.1 | 8 | 1177 | BAC29375.1 |
More Information
Set Membership: |
Intragenic Exonic |