Assay Details
Target Gene Details
Entrez Gene ID: | 240028 |
Gene Name: | leucyl/cystinyl aminopeptidase |
Gene Aliases: |
2010309L07Rik, 4732490P18Rik, CAP, IRAP, PLAP, gp160, vp165 |
Location: |
Chr.17:17527723-17624489 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lnpep | NM_172827.3 | NP_766415.1 | ||
AK029094.1 | BAC26293.1 | |||
AK036967.1 | BAC29651.1 | |||
BC066791.1 | AAH66791.1 | |||
BC120925.1 | ||||
BC120926.1 |
More Information
Set Membership: |
Intragenic Non-exonic |