Assay Details
Target Gene Details
Entrez Gene ID: | 211468 |
Gene Name: | potassium voltage-gated channel, subfamily H (eag-related), member 8 |
Gene Aliases: |
C130090D05Rik, ELK, ELK1, ELK3, Kv12.1 |
Location: |
Chr.17:52598233-52979864 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnh8 | NM_001031811.2 | 6 | 992 | NP_001026981.2 |
AK048629.1 | 6 | 881 | BAC33401.1 | |
BC029690.1 | 2 | 219 | AAH29690.1 |
More Information
Set Membership: |
Intragenic Exonic |