Assay Details
Target Gene Details
Entrez Gene ID: | 353130 |
Gene Name: | protease, serine 33 |
Gene Aliases: |
Eos, mT6, tryptase-6 |
Location: |
Chr.17:23833338-23836126 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss33 | NM_001081399.2 | NP_001074868.2 | ||
AY262280.1 | AAP20885.1 | |||
BC101950.1 | AAI01951.1 | |||
BC103541.1 | AAI03542.1 | |||
BC103542.1 | AAI03543.1 | |||
BQ893522.1 |
More Information
Set Membership: |
Intragenic Non-exonic |