Assay Details
Target Gene Details
Entrez Gene ID: | 17287 |
Gene Name: | meprin 1 alpha |
Gene Aliases: |
AI098089, AW107200, Mep-1, Mep-1a, Mep1 |
Location: |
Chr.17:43474324-43504144 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mep1a | NM_008585.2 | NP_032611.2 | ||
AK093914.1 | ||||
AK143906.1 | ||||
AK164734.1 | BAE37893.1 | |||
BC015258.1 | AAH15258.1 | |||
M74897.1 | AAA75354.1 |
More Information
Set Membership: |
Intragenic Non-exonic |