Assay Details
Target Gene Details
Entrez Gene ID: | 17287 |
Gene Name: | meprin 1 alpha |
Gene Aliases: |
AI098089, AW107200, Mep-1, Mep-1a, Mep1 |
Location: |
Chr.17:43474324-43504144 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mep1a | NM_008585.2 | 10 | 1046 | NP_032611.2 |
AK093914.1 | 10 | 1005 | ||
AK143906.1 | 10 | 1004 | ||
AK164734.1 | 10 | 1001 | BAE37893.1 | |
BC015258.1 | 10 | 1046 | AAH15258.1 | |
M74897.1 | 10 | 1045 | AAA75354.1 |
More Information
Set Membership: |
Intragenic Exonic |