Assay Details
Target Gene Details
Entrez Gene ID: | 76890 |
Gene Name: | mediator of cell motility 1 |
Gene Aliases: |
0610016J10Rik, D930048L02Rik |
Location: |
Chr.17:74198398-74295544 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Memo1 | NM_133771.2 | NP_598532.1 | ||
AF363447.1 | AAL34463.1 | |||
AK002639.1 | ||||
AK159560.1 | BAE35184.1 | |||
BC013819.1 | AAH13819.1 |