Assay Details
Target Gene Details
Entrez Gene ID: | 225010 |
Gene Name: | lysocardiolipin acyltransferase 1 |
Gene Aliases: |
AI181996, Agpat8, Alcat1, Gm91, Lycat |
Location: |
Chr.17:73107985-73243368 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lclat1 | NM_001081071.2 | NP_001074540.1 | ||
NM_001177967.1 | NP_001171438.1 | |||
NM_001177968.1 | NP_001171439.1 | |||
AK144580.1 | BAE25946.1 | |||
BC147499.1 | ||||
BC150884.1 | ||||
BC158076.1 | ||||
BC158079.1 |
More Information
Set Membership: |
Intragenic Non-exonic |