Assay Details
Target Gene Details
Entrez Gene ID: | 213556 |
Gene Name: | pleckstrin homology domain containing, family H (with MyTH4 domain) member 2 |
Gene Aliases: |
AI256725, E030001K05, mKIAA2028 |
Location: |
Chr.17:84511895-84622142 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plekhh2 | NM_177606.4 | 7 | 629 | NP_808274.2 |
AK029252.1 | 7 | 631 | BAC26356.1 | |
AK173337.1 | 4 | 285 | BAD32615.1 | |
BC021518.1 | 7 | 601 | AAH21518.1 | |
BC125583.1 | 7 | 594 | ||
BC137804.1 | 7 | 591 |