Assay Details
Target Gene Details
Entrez Gene ID: | 18754 |
Gene Name: | protein kinase C, epsilon |
Gene Aliases: |
5830406C15Rik, PKC[e], PKCepsilon, Pkce, R75156 |
Location: |
Chr.17:86166971-86657919 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prkce | NM_011104.3 | 6 | 1570 | NP_035234.1 |
AF028009.1 | 6 | 771 | AAB84189.1 | |
AF325507.1 | 6 | 762 | AAG53692.1 | |
AK082438.1 | 6 | 1571 | ||
AK085351.1 | 7 | 982 |
More Information
Set Membership: |
Intragenic Exonic |