Assay Details
Target Gene Details
Entrez Gene ID: | 72900 |
Gene Name: | NADH dehydrogenase (ubiquinone) flavoprotein 2 |
Gene Aliases: |
2900010C23Rik |
Location: |
Chr.17:66078795-66101559 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufv2 | NM_001278415.1 | 7 | 720 | NP_001265344.1 |
NM_028388.3 | 6 | 623 | NP_082664.1 | |
AK013511.1 | 6 | 574 | BAB28888.1 | |
AK078351.1 | 6 | 567 | BAC37233.1 | |
AK088193.1 | 3 | 362 | BAC40201.1 | |
AK146998.1 | 6 | 557 | BAE27595.1 | |
AK150404.1 | 7 | 657 | BAE29530.1 | |
AK151729.1 | 7 | 657 | BAE30647.1 | |
AK159460.1 | 6 | 540 | BAE35102.1 | |
AK166539.1 | 6 | 584 | BAE38841.1 | |
AK169143.1 | 6 | 594 | BAE40922.1 | |
AK185171.1 | 2 | 246 | ||
BC030946.1 | 6 | 552 | AAH30946.1 | |
CF106105.1 | 5 | 465 |
More Information
Set Membership: |
Intragenic Exonic |