Assay Details
Target Gene Details
Entrez Gene ID: | 213389 |
Gene Name: | PR domain containing 9 |
Gene Aliases: |
BC012016, Dsbc1, G1-419-29, Meisetz, Rcr1, repro7 |
Location: |
Chr.17:15543072-15564379 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prdm9 | NM_144809.2 | NP_659058.2 | ||
AB844106.1 | ||||
AB844107.1 | ||||
AB844108.1 | ||||
AB844109.1 | ||||
AB844110.1 | ||||
AB844111.1 | ||||
AB844112.1 | ||||
AB844113.1 | ||||
AB844114.1 | ||||
BC012016.1 | AAH12016.1 | |||
BC023014.1 | ||||
BC049903.1 | ||||
FJ212287.1 | ||||
GU216230.1 | ||||
HQ704390.1 | ||||
HQ704391.1 |
More Information
Set Membership: |
Intragenic Non-exonic |