Assay Details
Target Gene Details
Entrez Gene ID: | 68342 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10 |
Gene Aliases: |
0610011B04Rik, 22kDa, PDSW |
Location: |
Chr.17:24722067-24724388 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufb10 | NM_026684.2 | NP_080960.1 | ||
AK002517.1 | BAB22156.1 | |||
AK151404.1 | BAE30371.1 | |||
BC031664.1 | AAH31664.1 | |||
BC043013.1 | AAH43013.1 |
More Information
Set Membership: |
Intragenic Non-exonic |