Assay Details
Target Gene Details
Entrez Gene ID: | 104103 |
Gene Name: | antisense Igf2r RNA |
Gene Aliases: |
2810051F02Rik, 2810434M15Rik, AI256653, AI597500, AW049873, Air, B930018I07Rik, D17Ertd663e, IGF2RAS |
Location: |
Chr.17:12741311-12859884 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Airn | NR_027773.1 | |||
AK013242.1 | ||||
BY718621.1 | ||||
DQ275620.1 |
Target Gene Details
Entrez Gene ID: | 17171 |
Gene Name: | MAS1 oncogene |
Gene Aliases: |
Mas-1, Mgra |
Location: |
Chr.17:12838841-12868145 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mas1 | NM_008552.4 | 8 | 827 | NP_032578.2 |
AK015036.1 | 8 | 818 | BAB29684.1 | |
AK030261.1 | 8 | 817 | BAC26865.1 | |
AK030265.1 | 8 | 828 | BAC26868.1 | |
BC120790.1 | 3 | 149 | ||
BC125537.1 | 4 | 212 | ||
BC144900.1 | 3 | 161 | ||
U96273.1 | 3 | 223 | AAB69120.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |