Assay Details
Target Gene Details
Entrez Gene ID: | 50873 |
Gene Name: | Parkinson disease (autosomal recessive, juvenile) 2, parkin |
Gene Aliases: |
PRKN |
Location: |
Chr.17:10840384-12063370 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Park2 | NM_016694.3 | 11 | 1323 | NP_057903.1 |
AB019558.1 | 11 | 1294 | BAA82404.1 | |
AF250293.1 | 11 | 1323 | AAG13890.1 | |
AK015990.1 | 2 | 307 | BAB30069.1 | |
BC113204.1 | 11 | 1306 | AAI13205.1 | |
FJ555554.1 | 9 | 1076 | ||
FJ555555.1 | 6 | 734 | ||
FJ555556.1 | 10 | 1056 | ||
FJ555557.1 | 10 | 1181 | ||
GU272056.1 | 5 | 597 |