Assay Details
Target Gene Details
Entrez Gene ID: | 50873 |
Gene Name: | Parkinson disease (autosomal recessive, juvenile) 2, parkin |
Gene Aliases: |
PRKN |
Location: |
Chr.17:10840384-12063370 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Park2 | NM_016694.3 | NP_057903.1 | ||
AB019558.1 | BAA82404.1 | |||
AF250293.1 | AAG13890.1 | |||
BC113204.1 | AAI13205.1 | |||
FJ555554.1 | ||||
FJ555555.1 | ||||
FJ555556.1 | ||||
FJ555557.1 | ||||
GU272056.1 |