Assay Details
Target Gene Details
Entrez Gene ID: | 26416 |
Gene Name: | mitogen-activated protein kinase 14 |
Gene Aliases: |
CSBP2, Crk1, Csbp1, Mxi2, PRKM14, PRKM15, p38, p38-alpha, p38MAPK, p38a, p38alpha |
Location: |
Chr.17:28691342-28748405 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mapk14 | NM_001168508.1 | NP_001161980.1 | ||
NM_001168513.1 | NP_001161985.1 | |||
NM_001168514.1 | NP_001161986.1 | |||
NM_011951.3 | NP_036081.1 | |||
AF128892.1 | AAF34818.1 | |||
AF195850.1 | AAF06348.1 | |||
AK089059.1 | BAC40726.1 | |||
AK133684.1 | BAE21782.1 | |||
AK151348.1 | BAE30324.1 | |||
AK153025.1 | BAE31659.1 | |||
BC012235.1 | AAH12235.1 | |||
BC085303.1 | AAH85303.1 | |||
CX568915.1 | ||||
D83073.1 | BAA19741.1 | |||
U10871.1 | AAA20888.1 | |||
X65067.1 | CAA46200.1 |
More Information
Set Membership: |
Intragenic Non-exonic |