Assay Details
Target Gene Details
Entrez Gene ID: | 14651 |
Gene Name: | hydroxyacyl glutathione hydrolase |
Gene Aliases: |
BC019817, Glo-2, Glo2, Rsp29 |
Location: |
Chr.17:24840079-24864450 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hagh | NM_001159626.1 | 2 | 133 | NP_001153098.1 |
NM_024284.2 | 2 | 223 | NP_077246.2 | |
AK147022.1 | 2 | 184 | BAE27615.1 | |
AK150557.1 | 2 | 141 | BAE29657.1 | |
AK151231.1 | 2 | 141 | BAE30223.1 | |
AK152742.1 | 2 | 141 | BAE31462.1 | |
AK165777.1 | 2 | 127 | BAE38377.1 | |
BC004749.1 | 2 | 166 | AAH04749.1 | |
BC019817.1 | 2 | 116 | AAH19817.1 | |
BY348635.1 | 2 | 224 |