Assay Details
Target Gene Details
Entrez Gene ID: | 18763 |
Gene Name: | polycystic kidney disease 1 homolog |
Gene Aliases: |
PC1, mFLJ00285 |
Location: |
Chr.17:24549950-24596514 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pkd1 | NM_013630.2 | 7 | 1765 | NP_038658.2 |
AK220312.1 | 3 | 832 | BAD90231.1 | |
U70209.1 | 7 | 1767 | AAC53207.1 |
More Information
Set Membership: |
Intragenic Exonic |