Assay Details
Target Gene Details
Entrez Gene ID: | 16004 |
Gene Name: | insulin-like growth factor 2 receptor |
Gene Aliases: |
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300 |
Location: |
Chr.17:12682406-12769706 on Build GRCm38 |
Assay Gene Location: | Within Exon 41 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igf2r | NM_010515.2 | 41 | 6288 | NP_034645.2 |
AK053045.1 | 6 | 880 | BAC35249.1 | |
AK088699.1 | 16 | 2514 | BAC40512.1 | |
BC031190.1 | 8 | 1294 | AAH31190.1 | |
BC043457.1 | 4 | 489 | AAH43457.1 | |
BC055018.1 | 4 | 494 | AAH55018.1 | |
BC150817.1 | 41 | 6199 | ||
BC171973.1 | 41 | 6199 | ||
U04710.1 | 41 | 6244 | AAA19568.1 | |
X60389.1 | CAA42940.1 |
More Information
Set Membership: |
Intragenic Exonic |