Assay Details
Target Gene Details
Entrez Gene ID: | 353344 |
Gene Name: | opsin 5 |
Gene Aliases: |
Gpr136, Neuropsin, PGR12, TMEM13 |
Location: |
Chr.17:42556779-42620470 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Opn5 | NM_181753.4 | 5 | 799 | NP_861418.2 |
AK039525.1 | BAC30373.1 | |||
AY288426.1 | 5 | 786 | AAP72135.1 | |
AY318865.1 | 5 | 800 | AAR08201.1 | |
BC125611.1 | 5 | 881 | ||
BC132010.1 | 5 | 881 |
More Information
Set Membership: |
Intragenic Exonic |