Assay Details
Target Gene Details
Entrez Gene ID: | 67681 |
Gene Name: | mitochondrial ribosomal protein L18 |
Gene Aliases: |
1010001C05Rik, C79879, HSPC071, MRP-L18 |
Location: |
Chr.17:12911345-12916478 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrpl18 | NM_026310.3 | NP_080586.1 | ||
AK003104.1 | BAB22567.1 | |||
AK007465.1 | ||||
AK008680.1 | BAB25828.1 | |||
AK010150.1 | BAB26735.1 | |||
AK089039.1 | BAC40714.1 | |||
BC011425.1 | AAH11425.1 |
More Information
Set Membership: |
Intragenic Non-exonic |