Assay Details
Target Gene Details
Entrez Gene ID: | 107971 |
Gene Name: | fibroblast growth factor receptor substrate 3 |
Gene Aliases: |
4930417B13Rik, AI449674, Frs2beta, Snt2 |
Location: |
Chr.17:47687296-47704286 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Frs3 | NM_144939.2 | NP_659188.1 | ||
BC014819.1 | AAH14819.1 |
Target Gene Details
Entrez Gene ID: | 100126242 |
Gene Name: | fibroblast growth factor receptor substrate 3, opposite strand |
Gene Aliases: |
4930417B13Rik, Gm14873, OTTMUSG00000018358 |
Location: |
Chr.17:47699969-47701996 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Frs3os | NR_045912.1 | |||
NR_045913.1 | ||||
AK076701.1 |