Assay Details
Target Gene Details
Entrez Gene ID: | 104103 |
Gene Name: | antisense Igf2r RNA |
Gene Aliases: |
2810051F02Rik, 2810434M15Rik, AI256653, AI597500, AW049873, Air, B930018I07Rik, D17Ertd663e, IGF2RAS |
Location: |
Chr.17:12741311-12859884 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Airn | NR_002853.2 | |||
NR_027772.1 | ||||
NR_027773.1 | ||||
NR_027784.1 | ||||
AK012942.1 | ||||
AK013242.1 | ||||
AK032756.1 | 1 | 417 | ||
BF729958.1 | ||||
BI872620.1 | ||||
BY718621.1 | ||||
DQ275617.1 | ||||
DQ275619.1 | ||||
DQ275620.1 |
Target Gene Details
Entrez Gene ID: | 16004 |
Gene Name: | insulin-like growth factor 2 receptor |
Gene Aliases: |
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300 |
Location: |
Chr.17:12682406-12769706 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igf2r | NM_010515.2 | 2 | 380 | NP_034645.2 |
BB865604.1 | 2 | 330 | ||
BC150817.1 | 2 | 291 | ||
BC171973.1 | 2 | 291 | ||
FJ425888.1 | 2 | 187 | ||
FJ425889.1 | 2 | 202 | ||
U04710.1 | 2 | 336 | AAA19568.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |