Assay Details
Target Gene Details
Entrez Gene ID: | 76497 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 11 |
Gene Aliases: |
1500041B02Rik, AV117883, Hcgv, Tcte5, Tctex-5, Tctex5 |
Location: |
Chr.17:36948355-36951792 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppp1r11 | NM_029632.3 | NP_083908.1 | ||
AK005379.1 | BAB23986.1 | |||
AK077455.1 | ||||
AK089107.1 | BAC40754.1 | |||
AK162607.1 | ||||
AK170104.1 | BAE41566.1 | |||
BC027737.1 | AAH27737.1 | |||
EF528576.1 | ||||
EF528577.1 | ||||
EF528578.1 | ||||
EF528579.1 |
More Information
Set Membership: |
Intragenic Non-exonic |