Assay Details
Target Gene Details
Entrez Gene ID: | 378462 |
Gene Name: | MORN repeat containing 2 |
Gene Aliases: |
Mopt |
Location: |
Chr.17:80290212-80305691 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Morn2 | NM_194269.2 | NP_919245.1 | ||
AY367765.1 | AAQ73169.1 | |||
BC089486.1 | AAH89486.1 |