Assay Details
Target Gene Details
Entrez Gene ID: | 76438 |
Gene Name: | raftlin lipid raft linker 1 |
Gene Aliases: |
2310015N21Rik, AI853526 |
Location: |
Chr.17:49993258-50190497 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rftn1 | NM_181397.2 | 6 | 1219 | NP_852062.1 |
AK088466.1 | 4 | 756 | ||
AK152130.1 | 6 | 1141 | BAE30971.1 | |
AK153288.1 | 6 | 1141 | BAE31871.1 | |
AK155870.1 | 6 | 1183 | BAE33473.1 | |
AK170913.1 | 6 | 1182 | ||
AK172884.1 | 6 | 1147 | BAD32162.1 | |
BC043324.1 | 2 | 276 | AAH43324.1 | |
BC052074.1 | 6 | 1219 | AAH52074.1 |