Assay Details
Target Gene Details
Entrez Gene ID: | 76920 |
Gene Name: | arrestin domain containing 5 |
Gene Aliases: |
1700013E09Rik |
Location: |
Chr.17:56294111-56300286 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arrdc5 | NM_029799.1 | 2 | 358 | NP_084075.1 |
BC100496.1 | 2 | 358 | AAI00497.1 |