Assay Details
Target Gene Details
Entrez Gene ID: | 66968 |
Gene Name: | perilipin 5 |
Gene Aliases: |
2310076L09Rik, AI415325, AW109675, Lsdp5, MLDP, PAT-1 |
Location: |
Chr.17:56111597-56117606 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plin5 | NM_001077348.1 | 3 | 335 | NP_001070816.1 |
NM_025874.3 | 4 | 281 | NP_080150.2 | |
AK010202.1 | 4 | 281 | ||
AK075872.1 | 4 | 278 | BAC36019.1 | |
AY919875.1 | 3 | 294 | AAX12443.1 | |
BC024138.1 | 4 | 261 | ||
BY101690.1 | 4 | 282 | ||
DQ473305.1 | 4 | 261 |
More Information
Set Membership: |
Intragenic Exonic |