Assay Details
Target Gene Details
Entrez Gene ID: | 69542 |
Gene Name: | RIKEN cDNA 2300002M23 gene |
Gene Aliases: |
Stg, emprin |
Location: |
Chr.17:35567485-35568945 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
2300002M23Rik | NM_175148.3 | NP_780357.1 | ||
AK037046.1 | BAC29684.1 | |||
BC107265.1 | AAI07266.1 | |||
BC107266.1 | AAI07267.1 |