Assay Details
Target Gene Details
Entrez Gene ID: | 55979 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 1 (lysophosphatidic acid acyltransferase, alpha) |
Gene Aliases: |
1-AGP, 1-AGPAT, AW047140 |
Location: |
Chr.17:34604278-34613449 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agpat1 | NM_001163379.1 | 7 | 2127 | NP_001156851.1 |
NM_018862.3 | 7 | 1879 | NP_061350.3 | |
AK086281.1 | BAC39641.1 | |||
BC009651.1 | 7 | 1879 | AAH09651.1 |
Target Gene Details
Entrez Gene ID: | 81701 |
Gene Name: | EGF-like domain 8 |
Gene Aliases: |
Ng3 |
Location: |
Chr.17:34613200-34616572 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Egfl8 | NM_152922.3 | NP_690886.3 | ||
AK148723.1 | BAE28647.1 | |||
AY635583.1 | AAT47548.1 | |||
BC139484.1 | ||||
BC141312.1 | ||||
BC145632.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |