Assay Details
Target Gene Details
Entrez Gene ID: | 54419 |
Gene Name: | claudin 6 |
Gene Aliases: |
AL024037 |
Location: |
Chr.17:23679348-23682446 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cldn6 | NM_018777.4 | NP_061247.1 | ||
AK010560.1 | BAB27028.1 | |||
AK010682.1 | BAB27114.1 | |||
BC005718.1 | AAH05718.1 | |||
BC050138.1 | AAH50138.1 |
More Information
Set Membership: |
Intragenic Non-exonic |