Assay Details
Target Gene Details
Entrez Gene ID: | 52614 |
Gene Name: | adhesion G protein-coupled receptor E4 |
Gene Aliases: |
D17Ertd479e, Egf-tm7, Emr4, Fire, Gpr127 |
Location: |
Chr.17:55749984-55853662 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adgre4 | NM_139138.3 | NP_631877.2 | ||
AF396935.1 | AAL31879.1 | |||
AK036459.1 | ||||
AK037483.1 | BAC29816.1 | |||
AK155935.1 | BAE33510.1 | |||
AY032690.1 | AAK51125.1 | |||
BC120538.1 | ||||
BC120540.1 |
More Information
Set Membership: |
Intragenic Non-exonic |