Assay Details
Target Gene Details
Entrez Gene ID: | 224823 |
Gene Name: | ribosomal RNA processing 36 homolog (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.17:46667442-46674451 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rrp36 | NM_144857.1 | NP_659106.1 | ||
AK148237.1 | BAE28429.1 | |||
BC011248.1 | AAH11248.1 |