Assay Details
Target Gene Details
Entrez Gene ID: | 12263 |
Gene Name: | complement component 2 (within H-2S) |
Gene Aliases: |
- |
Location: |
Chr.17:34862602-34882100 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C2 | NM_013484.2 | NP_038512.2 | ||
AK146812.1 | BAE27452.1 | |||
AK194054.1 | ||||
AK211697.1 | ||||
BC011086.1 | AAH11086.1 | |||
M57891.1 | AAA63294.1 |