Assay Details
Target Gene Details
Entrez Gene ID: | 53817 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B |
Gene Aliases: |
0610030D10Rik, AI428441, Bat-1, Bat1, Bat1a, D17H6S81E, D17H6S81E-1, D6S81Eh |
Location: |
Chr.17:35241746-35253707 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx39b | NM_001252457.1 | 11 | 1554 | NP_001239386.1 |
NM_019693.3 | 11 | 1642 | NP_062667.1 | |
AA231260.1 | 2 | 212 | ||
AK051034.1 | 11 | 1575 | BAC34505.1 | |
AK088867.1 | 11 | 1560 | BAC40624.1 | |
AK196028.1 | 1 | 64 | ||
AK202733.1 | 2 | 203 | ||
BC011067.1 | 11 | 1550 | AAH11067.1 | |
BC024859.1 | 10 | 2778 | AAH24859.1 |