Assay Details
Target Gene Details
Entrez Gene ID: | 73242 |
Gene Name: | alpha tubulin acetyltransferase 1 |
Gene Aliases: |
0610011P08Rik, 2610008K08Rik, 2610110G12Rik, 3110080J08Rik, Mec17, TAT |
Location: |
Chr.17:35897595-35910093 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atat1 | NM_001142744.1 | NP_001136216.1 | ||
NM_001142745.1 | NP_001136217.1 | |||
NM_028476.4 | NP_082752.3 | |||
AK002569.1 | BAB22194.1 | |||
AK011350.1 | ||||
AK011750.2 | ||||
AK011838.1 | ||||
AK014265.1 | ||||
AK028297.1 | BAC25866.1 | |||
AK034725.1 | BAC28808.1 | |||
AK049682.1 | ||||
AK133694.1 | BAE21786.1 | |||
BB857404.1 | ||||
BC028847.1 | AAH28847.1 | |||
BC030669.1 | AAH30669.1 |