Assay Details
Target Gene Details
Entrez Gene ID: | 12305 |
Gene Name: | discoidin domain receptor family, member 1 |
Gene Aliases: |
6030432F18, AI323681, CD167a, Cak, Nep, PTK3A |
Location: |
Chr.17:35681567-35704629 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddr1 | NM_001198831.1 | 13 | 1787 | NP_001185760.1 |
NM_001198833.1 | 13 | 1906 | NP_001185762.1 | |
NM_007584.2 | 13 | 1962 | NP_031610.1 | |
NM_172962.1 | 12 | 1851 | NP_766550.1 | |
AF026259.1 | 13 | 1804 | AAB81866.1 | |
AK031442.1 | 12 | 1754 | BAC27407.1 | |
AK138698.1 | 13 | 1775 | ||
AK143088.1 | 12 | 1744 | BAE25267.1 | |
BC037108.1 | 14 | 2041 | ||
BC065998.1 | 12 | 1687 | AAH65998.1 | |
L57509.1 | 13 | 1810 | AAB05209.1 |
More Information
Set Membership: |
Intragenic Exonic |