Assay Details
Target Gene Details
Entrez Gene ID: | 68262 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 4 (lysophosphatidic acid acyltransferase, delta) |
Gene Aliases: |
1500003P24Rik |
Location: |
Chr.17:12118709-12219649 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agpat4 | NM_026644.2 | NP_080920.2 | ||
AK005139.1 | BAB23837.2 | |||
AK166917.1 | BAE39115.1 | |||
BC047281.1 | AAH47281.1 |
More Information
Set Membership: |
Intragenic Non-exonic |