Assay Details
Target Gene Details
Entrez Gene ID: | 55979 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 1 (lysophosphatidic acid acyltransferase, alpha) |
Gene Aliases: |
1-AGP, 1-AGPAT, AW047140 |
Location: |
Chr.17:34604278-34613449 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agpat1 | NM_001163379.1 | NP_001156851.1 | ||
NM_018862.3 | NP_061350.3 | |||
AB005623.1 | BAA22599.1 | |||
AK086281.1 | BAC39641.1 | |||
BC009651.1 | AAH09651.1 |
More Information
Set Membership: |
Intragenic Non-exonic |