Assay Details
Target Gene Details
Entrez Gene ID: | 73694 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 7 |
Gene Aliases: |
2410091C18Rik, AL033374, PRO1853 |
Location: |
Chr.17:78937106-78948052 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufaf7 | NM_028611.3 | NP_082887.2 | ||
AK010752.1 | BAB27158.2 | |||
AK090097.1 | BAC41089.1 | |||
AK184109.1 | ||||
BC028524.1 | 1 | 76 | AAH28524.1 | |
BC108350.1 | AAI08351.1 |