Assay Details
Target Gene Details
Entrez Gene ID: | 12703 |
Gene Name: | suppressor of cytokine signaling 1 |
Gene Aliases: |
Cish1, Cish7, JAB, SOCS-1, SSI-1 |
Location: |
Chr.16:10783808-10785536 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Socs1 | NM_001271603.1 | NP_001258532.1 | ||
NM_009896.2 | NP_034026.1 | |||
BC132366.1 | ||||
BC132368.1 |
More Information
Set Membership: |
Intragenic Non-exonic |