Assay Details
Target Gene Details
Entrez Gene ID: | 239759 |
Gene Name: | lipase, member H |
Gene Aliases: |
C130037N08Rik, D16Wsu119e, H06, Lpdlr, P3, PLA1B, lpd1, lpd2, mPA-PLA1 |
Location: |
Chr.16:21953817-21995542 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Liph | NM_001083894.1 | 3 | 655 | NP_001077363.1 |
NM_001289581.1 | 3 | 714 | NP_001276510.1 | |
NM_001289582.1 | 3 | 655 | NP_001276511.1 | |
NM_153404.3 | 3 | 655 | NP_700453.1 | |
AK035498.1 | 3 | 608 | ||
AK048150.1 | 3 | 554 | BAC33259.1 | |
AK134352.1 | 3 | 614 | BAE22108.1 | |
AK136503.1 | 3 | 618 | BAE23014.1 | |
AK137278.1 | 3 | 686 | ||
AK161067.1 | 3 | 599 | BAE36182.1 | |
AK162492.1 | 3 | 621 | BAE36944.1 | |
AY093499.1 | 3 | 482 | AAM18804.1 | |
BC037489.1 | 3 | 573 | AAH37489.1 |
More Information
Set Membership: |
Intragenic Exonic |