Assay Details
Target Gene Details
Entrez Gene ID: | 72058 |
Gene Name: | immunoglobulin superfamily, member 5 |
Gene Aliases: |
2010003D20Rik, JCAM, Jam4 |
Location: |
Chr.16:96360467-96465572 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igsf5 | NM_001177886.1 | NP_001171357.1 | ||
NM_001177887.1 | NP_001171358.1 | |||
NM_028078.3 | NP_082354.1 | |||
AF537215.1 | AAP49218.1 | |||
AK008060.1 | BAB25436.1 | |||
AK078934.1 | BAC37469.1 | |||
AK086973.1 | ||||
BC004806.1 | AAH04806.1 |
More Information
Set Membership: |
Intragenic Non-exonic |