Assay Details
Target Gene Details
Entrez Gene ID: | 67581 |
Gene Name: | TBC1 domain family, member 23 |
Gene Aliases: |
4930451A13Rik, AU015720, AU043671, AU043778, D030022P07Rik, W51689 |
Location: |
Chr.16:57168858-57231532 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tbc1d23 | NM_026254.2 | NP_080530.2 | ||
AK015437.1 | ||||
AK155142.1 | BAE33073.1 | |||
BC019762.1 | AAH19762.1 | |||
BC031706.1 | AAH31706.1 |