Assay Details
Target Gene Details
Entrez Gene ID: | 268902 |
Gene Name: | roundabout homolog 2 (Drosophila) |
Gene Aliases: |
2600013A04Rik, 9430089E08Rik, BB097918, D230004I22Rik, mKIAA1568 |
Location: |
Chr.16:73892061-75447298 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Robo2 | NM_175549.4 | NP_780758.3 | ||
AK035110.1 | ||||
AK085461.1 | ||||
AK129396.1 | BAC98206.1 | |||
BC055333.1 | AAH55333.1 | |||
DQ533875.1 | ||||
DQ533876.1 |