Assay Details
Target Gene Details
Entrez Gene ID: | 77626 |
Gene Name: | sphingomyelin phosphodiesterase 4 |
Gene Aliases: |
4122402O22Rik, mKIAA1418 |
Location: |
Chr.16:17619354-17644830 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smpd4 | NM_001164609.1 | 15 | 1437 | NP_001158081.1 |
NM_001164610.1 | 14 | 1398 | NP_001158082.1 | |
NM_001164611.1 | 14 | 1395 | NP_001158083.1 | |
NM_029945.3 | 15 | 1485 | NP_084221.2 | |
AK019466.2 | 15 | 1486 | BAB31737.1 | |
AK031029.1 | 14 | 1389 | BAC27221.1 | |
AK031685.1 | 14 | 1397 | BAC27512.1 | |
AK035892.1 | 14 | 1390 | BAC29229.1 | |
AK036055.1 | 14 | 1390 | ||
AK036380.1 | 14 | 1394 | ||
AK129354.1 | 15 | 1462 | BAC98164.1 | |
AK138720.1 | 14 | 1372 | ||
AK164234.1 | 15 | 1429 | BAE37694.1 | |
AK164456.1 | 12 | 1863 | BAE37796.1 | |
BC026767.1 | 14 | 1371 | AAH26767.1 |
More Information
Set Membership: |
Intragenic Exonic |