Assay Details
Target Gene Details
Entrez Gene ID: | 26559 |
Gene Name: | hormonally upregulated Neu-associated kinase |
Gene Aliases: |
Bstk1, Mak-v |
Location: |
Chr.16:90385797-90499555 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hunk | NM_015755.2 | 10 | 1446 | NP_056570.1 |
AF055919.2 | 10 | 1446 | AAC61489.1 | |
AF167987.1 | 10 | 1458 | AAF35282.1 | |
AK144639.1 | 10 | 1429 | BAE25983.1 | |
AK147425.1 | 10 | 1831 | ||
AK147734.1 | 10 | 1200 | BAE28103.1 | |
BC137788.1 | 10 | 1507 | ||
BC137790.1 | 10 | 1506 |
More Information
Set Membership: |
Intragenic Exonic |