Assay Details
Target Gene Details
Entrez Gene ID: | 13972 |
Gene Name: | guanine nucleotide binding protein (G protein), beta polypeptide 1-like |
Gene Aliases: |
ESTM55, Gm16314, Me49f07, OTTMUSG00000033458, Wdr14, Wdvcf |
Location: |
Chr.16:18498021-18566680 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnb1l | NM_001081682.2 | NP_001075151.1 | ||
NM_001285491.1 | NP_001272420.1 | |||
NM_001285493.1 | NP_001272422.1 | |||
NM_001285494.1 | NP_001272423.1 | |||
NM_023120.5 | NP_075609.2 | |||
NR_104347.1 | ||||
AF301595.1 | AAG44996.1 | |||
AK010489.1 | BAB26980.1 | |||
AK016952.1 | ||||
AK087954.1 | ||||
AK158210.1 | BAE34406.1 | |||
AK166195.1 | BAE38622.1 | |||
AK171253.1 | BAE42345.1 | |||
AK173227.1 | BAD32505.1 | |||
BB640704.1 | ||||
BC024635.1 | AAH24635.1 | |||
BY328275.1 |
More Information
Set Membership: |
Intragenic Non-exonic |